Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium monofluorophosphate | 10163-15-2 | 25 G
SDP

Catalog No. 5000407373
Encompass_Preferred

Disodium monofluorophosphate is a biochemical reagent used in life science research. It is a white to off-white solid that is stable under recommended storage conditions and is soluble in water.

  • Biochemical reagent for life science research
  • Solid appearance (white to off-white)
  • Stable under recommended storage conditions
  • Soluble in water (≥ 100 mg/mL)
  • Recommended storage at 4°C, sealed, away from moisture
  • Suitable for laboratory chemicals and substance manufacturing

Catalog No. 50-004-07373 Supplier Medchemexpress LLC Supplier No. 25GHYB0540
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium monofluorophosphate is a biochemical reagent used in life science research. It is a white to off-white solid that is stable under recommended storage conditions and is soluble in water.

  • Biochemical reagent for life science research
  • Solid appearance (white to off-white)
  • Stable under recommended storage conditions
  • Soluble in water (≥ 100 mg/mL)
  • Recommended storage at 4°C, sealed, away from moisture
  • Suitable for laboratory chemicals and substance manufacturing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.