Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Prednisolone 21-Phosphate 98.0+%, TCI America™
SDP

Catalog No. D39185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39185G Supplier TCI America Supplier No. D39185G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125-02-0
Molecular Formula C21H27Na2O8P
Molecular Weight (g/mol) 484.392
MDL Number MFCD00083682
InChI Key VJZLQIPZNBPASX-ZWGKCJGLSA-L
Synonym Prednisolone 21-Phosphate Disodium Salt
PubChem CID 133109138
IUPAC Name disodium;[2-[(10S,13R,17S)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
SMILES CC12CC(C3C(C1CCC2(C(=O)COP(=O)([O-])[O-])O)CCC4=CC(=O)C=CC34C)O.[Na+].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 484.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Prednisolone 21-Phosphate
RTECSNumber TU4153300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.