Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Sebacate 97.0+%, TCI America™
SDP

Catalog No. S0270500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0270500G 500 g
S027025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0270500G Supplier TCI America Supplier No. S0270500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17265-14-4
Molecular Formula C10H16Na2O4
Molecular Weight (g/mol) 246.214
MDL Number MFCD00070501
InChI Key NCXUIEDQTCQZRK-UHFFFAOYSA-L
Synonym Decanedioic Acid Disodium Salt, Sebacic Acid Disodium Salt, Disodium Decanedioate
PubChem CID 134734
IUPAC Name disodium;decanedioate
SMILES C(CCCCC(=O)[O-])CCCC(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 246.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Sebacate
RTECSNumber VS1430000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.