Learn More
Medchemexpress LLC HY-W015410 100mg Medchemexpress, Disodium succinate CAS:150-90-3 Purity:>98%
SDP
Catalog No. 501970709
Encompass_Preferred
Medchemexpress, HY-W015410 100mg Disodium succinate CAS:150-90-3 Disodium succinate is the disodium salt of Succinic acid. Succinic acid is an intermediate product of the tricarboxylic acid cycle, as well as one of fermentation products of anaerobic metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0709 Supplier Medchemexpress LLC Supplier No. HYW015410100MG
Medchemexpress, HY-W015410 100mg Disodium succinate CAS:150-90-3 Disodium succinate is the disodium salt of Succinic acid. Succinic acid is an intermediate product of the tricarboxylic acid cycle, as well as one of fermentation products of anaerobic metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.