Learn More
Medchemexpress LLC Disodium succinate-13C2 | 83587-80-8 | 99.3% | 164.04 | C2 13C2H4Na2O4 | 100 MG
SDP
Catalog No. 5000303702
Encompass_Preferred
Disodium succinate-13C2 is the 13C-labeled isotopologue of disodium succinate supplied as a high-purity powder for use as an internal standard or tracer in metabolic and biochemical studies, particularly those involving the tricarboxylic acid (TCA) cycle. The compound is intended for analytical and research use in small milligram quantities and requires refrigerated, moisture-free storage.
  • 13C2 isotopic labeling suitable for tracer and metabolic studies.
  • High purity (99.3%) for reliable analytical results.
  • Available in multiple small pack sizes (5-100 mg) to match experimental needs.
  • Powder form; store at 4°C and keep sealed away from moisture.
  • Appropriate as an internal standard in mass spectrometry and metabolic assays.
Catalog No. 50-003-03702 Supplier Medchemexpress LLC Supplier No. HYW015410S100MG
Disodium succinate-13C2 is the 13C-labeled isotopologue of disodium succinate supplied as a high-purity powder for use as an internal standard or tracer in metabolic and biochemical studies, particularly those involving the tricarboxylic acid (TCA) cycle. The compound is intended for analytical and research use in small milligram quantities and requires refrigerated, moisture-free storage.
  • 13C2 isotopic labeling suitable for tracer and metabolic studies.
  • High purity (99.3%) for reliable analytical results.
  • Available in multiple small pack sizes (5-100 mg) to match experimental needs.
  • Powder form; store at 4°C and keep sealed away from moisture.
  • Appropriate as an internal standard in mass spectrometry and metabolic assays.