Learn More
Medchemexpress LLC Disodium succinate-13C2 | 83587-80-8 | 99.3% | 164.04 g/mol | C4H4Na2O4 | 5 MG
SDP
Catalog No. 5000303751
Encompass_Preferred
Disodium succinate-13C2 is the 13C2-stable-isotope labeled disodium salt of succinic acid supplied as a small-pack analytical standard for metabolic and biochemical research. It is used as an internal standard or tracer in mass spectrometry, NMR, and isotope-labeling experiments where high isotopic and chemical integrity are required.
  • 13C2 stable isotope labeling for tracing and quantification.
  • High chemical purity (reported 99.3%) for reliable analytical measurements.
  • Small pack size suitable for micro-scale analytical workflows.
  • Compatible with mass spectrometry and NMR applications.
  • Provides a defined reagent for metabolic flux and isotope tracing studies.
Catalog No. 50-003-03751 Supplier Medchemexpress LLC Supplier No. HYW015410S5MG
Disodium succinate-13C2 is the 13C2-stable-isotope labeled disodium salt of succinic acid supplied as a small-pack analytical standard for metabolic and biochemical research. It is used as an internal standard or tracer in mass spectrometry, NMR, and isotope-labeling experiments where high isotopic and chemical integrity are required.
  • 13C2 stable isotope labeling for tracing and quantification.
  • High chemical purity (reported 99.3%) for reliable analytical measurements.
  • Small pack size suitable for micro-scale analytical workflows.
  • Compatible with mass spectrometry and NMR applications.
  • Provides a defined reagent for metabolic flux and isotope tracing studies.