Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium succinate | 150-90-3 | MFCD00002790 | ≥98.0% | 162.05 g/mol | C4H4Na2O4 | 1 ML
SDP

Catalog No. 5000303267
Encompass_Preferred

Disodium succinate (CAS 150-90-3) is the disodium salt of succinic acid used as an intermediate of the tricarboxylic acid cycle and as a reagent, buffer component, or additive in chemical, pharmaceutical, food, and biochemical research. It is highly water-soluble and can be supplied as solid material or as prepared aqueous solutions for immediate use.

  • High water solubility (≥100 mg/mL) enables concentrated stock solutions.
  • Reliable molecular identity: C4H4Na2O4; molecular weight 162.05 g/mol.
  • Suitable as a buffering agent, additive, or reagent in biochemical assays.
  • Ready-to-use concentration examples simplify experimental setup.

Catalog No. 50-003-03267 Supplier Medchemexpress LLC Supplier No. HYW01541010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium succinate (CAS 150-90-3) is the disodium salt of succinic acid used as an intermediate of the tricarboxylic acid cycle and as a reagent, buffer component, or additive in chemical, pharmaceutical, food, and biochemical research. It is highly water-soluble and can be supplied as solid material or as prepared aqueous solutions for immediate use.

  • High water solubility (≥100 mg/mL) enables concentrated stock solutions.
  • Reliable molecular identity: C4H4Na2O4; molecular weight 162.05 g/mol.
  • Suitable as a buffering agent, additive, or reagent in biochemical assays.
  • Ready-to-use concentration examples simplify experimental setup.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.