Learn More
Medchemexpress LLC Disodium succinate | 150-90-3 | MFCD00002790 | 98.0% | 162.05 g/mol | C4H4Na2O4 | 250 MG
SDP
Catalog No. 5000303729
Encompass_Preferred
Disodium succinate is the disodium salt of succinic acid (CAS 150-90-3) provided for research and analytical use. It functions as a water-soluble platform chemical used across chemical, pharmaceutical, and food applications, and is commonly employed in buffering, flavoring, and biochemical studies.
  • High purity: 98.0%.
  • Water soluble (≥100 mg/mL) for preparing aqueous stock solutions.
  • Low solubility in DMSO (<1 mg/mL); choose solvent accordingly.
  • Functions as a buffer, chelating agent, surfactant, and flavoring additive.
  • Suitable for biochemical and analytical research applications.
Catalog No. 50-003-03729 Supplier Medchemexpress LLC Supplier No. 250MGHYW015410
Disodium succinate is the disodium salt of succinic acid (CAS 150-90-3) provided for research and analytical use. It functions as a water-soluble platform chemical used across chemical, pharmaceutical, and food applications, and is commonly employed in buffering, flavoring, and biochemical studies.
  • High purity: 98.0%.
  • Water soluble (≥100 mg/mL) for preparing aqueous stock solutions.
  • Low solubility in DMSO (<1 mg/mL); choose solvent accordingly.
  • Functions as a buffer, chelating agent, surfactant, and flavoring additive.
  • Suitable for biochemical and analytical research applications.