Learn More
Medchemexpress LLC Disodium succinate | 150-90-3 | MFCD00002790 | ≥98.0% | 162.05 g/mol | C4H4Na2O4 | 5 G
SDP
Catalog No. 5000302787
Encompass_Preferred
Disodium succinate is the disodium salt of succinic acid supplied as a white to off-white solid for laboratory and research use. It functions as a biochemical reagent, buffer component, flavoring agent, and endogenous metabolite in biochemical studies. The material is characterized by high purity and defined physicochemical properties suitable for analytical and preparative applications.
  • High purity (≥98%).
  • White to off-white solid form.
  • Molecular weight 162.05 g/mol.
  • Water-soluble for buffer and solution preparation.
  • Used as an endogenous metabolite in biochemical studies.
  • Applicable as a food additive and platform chemical.
Catalog No. 50-003-02787 Supplier Medchemexpress LLC Supplier No. HYW0154105G
Disodium succinate is the disodium salt of succinic acid supplied as a white to off-white solid for laboratory and research use. It functions as a biochemical reagent, buffer component, flavoring agent, and endogenous metabolite in biochemical studies. The material is characterized by high purity and defined physicochemical properties suitable for analytical and preparative applications.
  • High purity (≥98%).
  • White to off-white solid form.
  • Molecular weight 162.05 g/mol.
  • Water-soluble for buffer and solution preparation.
  • Used as an endogenous metabolite in biochemical studies.
  • Applicable as a food additive and platform chemical.