Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium succinate | 150-90-3 | MFCD00002790 | ≥98.0% | 162.05 g/mol | C4H4Na2O4 | 500 MG
SDP

Catalog No. 5000303269
Encompass_Preferred

Disodium succinate is the disodium salt of succinic acid supplied as a white to off-white solid for research and formulation applications. It serves as an endogenous metabolite and platform chemical for biochemical studies and food additive research, provided with supporting documentation for characterization and safe handling.

  • Purity ≥98.0%.
  • White to off-white solid.
  • Molecular formula C4H4Na2O4 and molecular weight 162.05 g/mol.
  • Soluble in water and DMSO.
  • Suitable for use in biochemical assays and metabolic studies.
  • Supplied with data sheet and safety data sheet for handling and storage.

Catalog No. 50-003-03269 Supplier Medchemexpress LLC Supplier No. HYW015410500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium succinate is the disodium salt of succinic acid supplied as a white to off-white solid for research and formulation applications. It serves as an endogenous metabolite and platform chemical for biochemical studies and food additive research, provided with supporting documentation for characterization and safe handling.

  • Purity ≥98.0%.
  • White to off-white solid.
  • Molecular formula C4H4Na2O4 and molecular weight 162.05 g/mol.
  • Soluble in water and DMSO.
  • Suitable for use in biochemical assays and metabolic studies.
  • Supplied with data sheet and safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.