Learn More
Medchemexpress LLC Disodium succinate | 83587-80-8 | 99.3% | 164.04 | C213C2H4Na2O4 | 50 MG
SDP
Catalog No. 5000302508
Encompass_Preferred
Disodium succinate-13C2 is the 13C2 isotopically labeled disodium salt of succinic acid, offered as a high-purity standard for analytical and biochemical use. It is intended for applications such as mass spectrometry, metabolic tracing, and quantitative assays where an isotopically labeled internal standard is required.
  • 13C2-labeled disodium succinate for isotopic tracing
  • High chemical purity (99.3%)
  • Suitable as an internal standard for mass spectrometry
  • Available in multiple milligram-scale package sizes
  • Stable, easy-to-handle solid form
Catalog No. 50-003-02508 Supplier Medchemexpress LLC Supplier No. HYW015410S50MG
Disodium succinate-13C2 is the 13C2 isotopically labeled disodium salt of succinic acid, offered as a high-purity standard for analytical and biochemical use. It is intended for applications such as mass spectrometry, metabolic tracing, and quantitative assays where an isotopically labeled internal standard is required.
  • 13C2-labeled disodium succinate for isotopic tracing
  • High chemical purity (99.3%)
  • Suitable as an internal standard for mass spectrometry
  • Available in multiple milligram-scale package sizes
  • Stable, easy-to-handle solid form