Learn More
Medchemexpress LLC Disodium succinate h 25g
SDP
Catalog No. 5000206371
Encompass_Preferred
Disodium succinate hexahydrate (Wormwood acid disodium hexahydrate) is an anti-anxiety agent with oral activity Disodium succinate hexahydrate is an intermediate product of the tricarboxylic acid cycle Disodium succinate hexahydrate is an important platform chemical Disodium succinate hexahydrate can be used as surfactant additive ion chelating agent flavoring agent and other applications in chemical pharmaceutical and food fields[1][2][3][4][5]
Catalog No. 50-002-06371 Supplier Medchemexpress LLC Supplier No. 25GHYW094516
Disodium succinate hexahydrate (Wormwood acid disodium hexahydrate) is an anti-anxiety agent with oral activity Disodium succinate hexahydrate is an intermediate product of the tricarboxylic acid cycle Disodium succinate hexahydrate is an important platform chemical Disodium succinate hexahydrate can be used as surfactant additive ion chelating agent flavoring agent and other applications in chemical pharmaceutical and food fields[1][2][3][4][5]