Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium terephthalate | 10028-70-3 | MFCD00013137 | 99.9% | 210.09 g/mol | C8H4Na2O4 | 25 G
SDP

Catalog No. 5000280592
Encompass_Preferred

Disodium terephthalate (terephthalic acid disodium salt) is a white to off-white solid used as a biochemical reagent and as a precursor in polyester (PET) synthesis. It is supplied for research use only.

  • High purity: 99.93%.
  • White to off-white solid appearance.
  • Molecular formula C8H4Na2O4; molecular weight 210.09 g/mol.
  • Suitable for biochemical assays and polymer precursor applications.
  • Storage: store at room temperature, keep dry and cool; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Available in multiple pack sizes including 25 G.

Catalog No. 50-002-80592 Supplier Medchemexpress LLC Supplier No. HYW25012725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium terephthalate (terephthalic acid disodium salt) is a white to off-white solid used as a biochemical reagent and as a precursor in polyester (PET) synthesis. It is supplied for research use only.

  • High purity: 99.93%.
  • White to off-white solid appearance.
  • Molecular formula C8H4Na2O4; molecular weight 210.09 g/mol.
  • Suitable for biochemical assays and polymer precursor applications.
  • Storage: store at room temperature, keep dry and cool; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Available in multiple pack sizes including 25 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.