Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Tetracarbonylferrate, Compound with 1,4-dioxane (1 to 1.5) 94%, Thermo Scientific™

Catalog No. AC209310100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC209310100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC209310100 Supplier Thermo Scientific Chemicals Supplier No. 209310100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59733-73-2
Molecular Formula C20H24Fe2Na4O14+4
Molecular Weight (g/mol) 692.047
InChI Key GNQHSDPSRBMMNX-UHFFFAOYSA-N
Synonym collman's reagent, collman inverted exclamation mark s reagent, disodium tetracarbonylferrate dioxane complex, tetrakis sodiomethylidyne oxidanium tetrakis carbon monoxide tris dioxane bis ?? 2-irondiuide
PubChem CID 71310151
IUPAC Name tetrasodium;carbon monoxide;1,4-dioxane;iron
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1COCCO1.C1COCCO1.C1COCCO1.[Na+].[Na+].[Na+].[Na+].[Fe].[Fe]

Specifications

Color Beige to Yellow or Orange
Quantity 10 g
Infrared Spectrum Authentic
Physical Form Powder
Formula Weight 346.02
Percent Purity ≥92%
Chemical Name or Material Disodium tetracarbonylferrate, 94%
Hazard Category GHS Signal Word: Danger
Hazard Statement AIR SENSITIVE
EINECSNumber 261-892-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.