Learn More
Medchemexpress LLC Disopyramide | 3737-09-5 | MFCD00057366 | 99.7% | 339.47 g/mol | C21H29N3O | 10MM 1ML
SDP
Catalog No. 5000237216
Encompass_Preferred
Disopyramide is a class IA antiarrhythmic agent used in research to investigate ventricular and atrial arrhythmias. It blocks fast inward sodium currents, prolongs cardiac action potential duration, and inhibits hERG-encoded potassium channels, making it useful for electrophysiology and pharmacology studies. Supplied for in vitro use with guidance for solvent preparation and storage.
Catalog No. 50-002-37216 Supplier Medchemexpress LLC Supplier No. HY1253310MM 1ML
Disopyramide is a class IA antiarrhythmic agent used in research to investigate ventricular and atrial arrhythmias. It blocks fast inward sodium currents, prolongs cardiac action potential duration, and inhibits hERG-encoded potassium channels, making it useful for electrophysiology and pharmacology studies. Supplied for in vitro use with guidance for solvent preparation and storage.