Learn More
Medchemexpress LLC Disopyramide phosphate | 22059-60-5 | MFCD00069254 | 99.9% | 437.47 | C21H32N3O5P | 100MG
SDP
Catalog No. 5000235673
Encompass_Preferred
Disopyramide phosphate is the phosphate salt of disopyramide, a class IA antiarrhythmic used in research to study cardiac electrophysiology. It blocks the fast inward sodium current, prolongs cardiac action potentials, inhibits hERG-encoded potassium channels, and exhibits negative inotropic effects. Supplied as a high-purity reference standard, it is intended for in vitro pharmacology and electrophysiology applications.
  • High purity suitable for analytical and reference use.
  • Available in multiple pack sizes including 100 mg for experimental flexibility.
  • Chemical formula C21H32N3O5P and molecular weight 437.47 g/mol for accurate dosing and calculations.
  • Relevant for sodium channel and cardiac action potential research.
  • Solid form enables straightforward storage and handling.
Catalog No. 50-002-35673 Supplier Medchemexpress LLC Supplier No. HY12533A100MG
Disopyramide phosphate is the phosphate salt of disopyramide, a class IA antiarrhythmic used in research to study cardiac electrophysiology. It blocks the fast inward sodium current, prolongs cardiac action potentials, inhibits hERG-encoded potassium channels, and exhibits negative inotropic effects. Supplied as a high-purity reference standard, it is intended for in vitro pharmacology and electrophysiology applications.
  • High purity suitable for analytical and reference use.
  • Available in multiple pack sizes including 100 mg for experimental flexibility.
  • Chemical formula C21H32N3O5P and molecular weight 437.47 g/mol for accurate dosing and calculations.
  • Relevant for sodium channel and cardiac action potential research.
  • Solid form enables straightforward storage and handling.