Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disopyramide 98.0+%, TCI America™
SDP

Catalog No. D27935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27935G 5 g
D27931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27935G Supplier TCI America Supplier No. D27935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9-5-3737 | C21H29N3O | 339.48 g/mol

Chemical Identifiers

CAS 9-5-3737
Molecular Formula C21H29N3O
Molecular Weight (g/mol) 339.48
MDL Number MFCD00057366
InChI Key UVTNFZQICZKOEM-UHFFFAOYNA-N
Synonym disopyramide, dicorantil, rythmodan, disopiramida, ritmodan, disopyramidum, searle 703, isorythm, lispine, disopyramidum inn-latin
PubChem CID 3114
ChEBI CHEBI:4657
IUPAC Name 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
SMILES CC(C)N(CCC(C1=CC=CC=C1)(C2=CC=CC=N2)C(=O)N)C(C)C

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 339.48
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Disopyramide
RTECSNumber UR8432000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.