Learn More
TWD Scientific LLC Small Antistatic Polypropylene Weighing Funnel. Pack of 50.
SDP
Catalog No. NC1900939 Shop All TWD Scientific LLC Products
Encompass_Preferred
Small disposable antistatic polypropylene weighing funnel. Fits volumetric flask volumes (ml) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL. 50/Pack
Catalog No. NC1900939 Supplier TWD Scientific LLC Supplier No. ASWF1S
Small disposable antistatic polypropylene weighing funnel. Fits volumetric flask volumes (ml) of 5, 10, 15, and 20. Funnels have a maximum capacity of 2mL. 50/Pack