Learn More
Bellco Glass, Inc. Disposible Baffle Shake Flask, 125mL Polycarbonate - Sterile
SDP
Catalog No. 501945614
Encompass
Sterile disposable Erlenmeyer culture flasks are supplied with 2-polypropylene closures, one with a solid top closure attached and another with a 0.2 micron PTFE vented closure. The flasks are graduated, non-pyrogenic and manufactured with Lexan virgin polycarbonate resin - non-pyrogenic.capacity: 125mldimensions: 67mm od x 111mm overall height3 side bafflesdisposable, re-autoclavable & re-freezablegraduated: yesneck size: 38-430supply with: two closures, solid top & .2um vented
Catalog No. 50-194-5614 Supplier Bellco Glass, Inc. Supplier No. 445520125
Sterile disposable Erlenmeyer culture flasks are supplied with 2-polypropylene closures, one with a solid top closure attached and another with a 0.2 micron PTFE vented closure. The flasks are graduated, non-pyrogenic and manufactured with Lexan virgin polycarbonate resin - non-pyrogenic.capacity: 125mldimensions: 67mm od x 111mm overall height3 side bafflesdisposable, re-autoclavable & re-freezablegraduated: yesneck size: 38-430supply with: two closures, solid top & .2um vented