Learn More
Biopac Systems Inc Disposable Cloth Facial Electrodes 60/pk
SDP
Catalog No. NC1568842
Encompass
Disposable cloth electrodes designed for recording EMG or ECG for sleep and facial applications. 10 mm contact area on 2 cm x 2 cm backing. Front has standard snap for lead connection. Back has conductive adhesive solid gel that tolerates repositioning for proper placement. The non-woven cloth base of the electrode is extremely conforming to contours of the face and very comfortable.
Catalog No. NC1568842 Supplier Biopac Systems Inc Supplier No. EL513
Disposable cloth electrodes designed for recording EMG or ECG for sleep and facial applications. 10 mm contact area on 2 cm x 2 cm backing. Front has standard snap for lead connection. Back has conductive adhesive solid gel that tolerates repositioning for proper placement. The non-woven cloth base of the electrode is extremely conforming to contours of the face and very comfortable.