Learn More
Bellco Glass, Inc. Disposible Cotton Plug/Past Pip,9in Borosilicate Glass
SDP
Catalog No. 501944652
Encompass_Preferred
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. MADE IN THE USA Case qty-1000
Catalog No. 50-194-4652 Supplier Bellco Glass, Inc. Supplier No. 127810009
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. MADE IN THE USA Case qty-1000