Learn More
Bellco Glass, Inc. Disp Culture Tube,25X100mm No Logo
SDP
Catalog No. 501945285
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USASpecifications:Flint Glass #2 Wall WeightType 1 Class A Borosilicate GlassSizes: 25x100mmClosure Size: 25mmCase Quantity: 144
Catalog No. 50-194-5285 Supplier Bellco Glass, Inc. Supplier No. 201025100
Add to Cart
Add to Cart
Manufactured from Type 1 Class A Borosilicate Glass. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USASpecifications:Flint Glass #2 Wall WeightType 1 Class A Borosilicate GlassSizes: 25x100mmClosure Size: 25mmCase Quantity: 144