Learn More
Bellco Glass, Inc. Disp Culture Tube, 25x150 mm
SDP
Catalog No. 501945286
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USASpecifications:Flint Glass #2 Wall WeightType 1 Class A Borosilicate GlassSizes: 25x150mmClosure Size: 25mmCase Quantity: 144
Catalog No. 50-194-5286 Supplier Bellco Glass, Inc. Supplier No. 201025150
Manufactured from Type 1 Class A Borosilicate Glass. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USASpecifications:Flint Glass #2 Wall WeightType 1 Class A Borosilicate GlassSizes: 25x150mmClosure Size: 25mmCase Quantity: 144