Learn More
Cole-Parmer Reinforced Nylon Disposable Serrated Round Tip Forceps, 4.9" L; 100/Pk
Catalog No. NC9655622 Shop All Cole-Parmer Products
Encompass_Preferred
White forceps feature corrugated handles and serrated blades. Rounded tips are useful when handling fragile samples. Autoclavable.
Catalog No. NC9655622 Supplier Cole-Parmer Supplier No. 0644315
White forceps feature corrugated handles and serrated blades. Rounded tips are useful when handling fragile samples. Autoclavable.