Learn More
Research Products International Corp Glass Pasteur Pipets
SDP
Catalog No. 502388407
Encompass
High quality borosilicate glass Pasteur pipets are ideal for inoculating culture media and transferring samples and reagents. Pipets can be used with aspirating bulbs or attached to a vacuum apparatus.
Catalog No. 50-238-8407 Supplier Research Products International Corp Supplier No. 248612A
High quality borosilicate glass Pasteur pipets are ideal for inoculating culture media and transferring samples and reagents. Pipets can be used with aspirating bulbs or attached to a vacuum apparatus.