Learn More
Azzota Corp Disposable Level 2 Isolation Gowns, Coat Type, Blue, 100/Pk
Catalog No. 502060398
Encompass_Preferred
DISPOSABLE Lightweight Multi-Ply Isolation Gown. Blue Color, Level 2 protection. Made of Non-woven Fabric. Soft and breathable for maximum comfort. Isolation gowns with level 2 protection are ideal for low-risk circumstances like blood draws from veins or work in pathology labs and ICUs and don’t require sterile gowns. Level 2 gowns are tested by pressurizing the material used to make the gowns and by impacting the gowns with water. These gowns can block more fluid than level 1 gowns and are effective against fluid penetration that might occur through splatter or soaking.
Catalog No. 50-206-0398 Supplier Azzota Corp Supplier No. GOWNL2100
DISPOSABLE Lightweight Multi-Ply Isolation Gown. Blue Color, Level 2 protection. Made of Non-woven Fabric. Soft and breathable for maximum comfort. Isolation gowns with level 2 protection are ideal for low-risk circumstances like blood draws from veins or work in pathology labs and ICUs and don’t require sterile gowns. Level 2 gowns are tested by pressurizing the material used to make the gowns and by impacting the gowns with water. These gowns can block more fluid than level 1 gowns and are effective against fluid penetration that might occur through splatter or soaking.