Learn More
Andwin Scientific DISP LIQ.TRANS PIPETTER 50/PK
SDP
Supplier:  Andwin Scientific 8889010818
Encompass_Preferred
The 12 mL transfer syringe is the most ideal pipette available as it can be used for several purposes, including dispensing epoxies, adhesives, lubricants, paints, oils, fluids, etc. This product can be used more than once or can be disposed right away. It is sterile and is individually wrapped. This is sold in a pack of 50.
Catalog No. NC9458448