Learn More
Waters Corp Disposable MicroCuvette Kit, 30 Pack, Cyclic Olefin Copolymers, 4 μL Minimum Volume
Catalog No. NC0616299 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This disposable cuvette kit provides 30 cuvettes and 3 reusable caps for use with the NanoStar I system. Made from cyclic olefin copolymers, these cuvettes are designed for aqueous solvents and have an operating temperature range of -10°C to 80°C.
  • Package of 30 cuvettes with 3 reusable caps.
  • Compatible with NanoStar I and DynaPro NanoStar II systems.
  • Made from cyclic olefin copolymers (COC).
  • Requires a minimum sample volume of 4μL.
  • Compatible with aqueous solvents only.
  • Operating temperature range: -10°C to 80°C.
Catalog No. NC0616299 Supplier Waters Corp Supplier No. WNDMC
This disposable cuvette kit provides 30 cuvettes and 3 reusable caps for use with the NanoStar I system. Made from cyclic olefin copolymers, these cuvettes are designed for aqueous solvents and have an operating temperature range of -10°C to 80°C.
  • Package of 30 cuvettes with 3 reusable caps.
  • Compatible with NanoStar I and DynaPro NanoStar II systems.
  • Made from cyclic olefin copolymers (COC).
  • Requires a minimum sample volume of 4μL.
  • Compatible with aqueous solvents only.
  • Operating temperature range: -10°C to 80°C.