Learn More
Bellco Glass, Inc. Disposable Pasteur Pipette 9In Soda Lime
SDP
Catalog No. 501944650
Encompass_Preferred
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand
Catalog No. 50-194-4650 Supplier Bellco Glass, Inc. Supplier No. 127720009
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand