Learn More
Bellco Glass, Inc. Disposible Pasteur Pipet,9in Borosilicate Glass
SDP
Catalog No. 501944648
Encompass_Preferred
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. MADE IN THE USA Case qty-1000
Catalog No. 50-194-4648 Supplier Bellco Glass, Inc. Supplier No. 127710009
For transfer of all infectious materials culture media, and routine sampling. Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. MADE IN THE USA Case qty-1000