Learn More
Andwin Scientific DISP PASTEUR PIPETTES 250/PK
SDP
Catalog No. 50930562
Encompass_Preferred
The disposable pasteur pipettes are ideal for handling infectious materials, inoculating culture media and routine sampling. This product has a body constructed with borosilicate Glass, an outside diameter of 7 mm, a length of 89 mm, and a tip length of 57 mm. It is solld in a pack of 250.
Catalog No. 50-930-562 Supplier Andwin Scientific Supplier No. 7095012
The disposable pasteur pipettes are ideal for handling infectious materials, inoculating culture media and routine sampling. This product has a body constructed with borosilicate Glass, an outside diameter of 7 mm, a length of 89 mm, and a tip length of 57 mm. It is solld in a pack of 250.