Learn More
MSC Guardair Disposable Drum Liners, 20 Gallon, 8 mil LDPE, 5 Pack
Catalog No. 502836338
Encompass
These disposable drum liners are designed for efficient recovery of wet or dry product from vacuum drums. Made from 8 mil low-density polyethylene, they feature heat-sealed seams and a rounded bottom for a strong, contoured, and wrinkle-free fit. The material is FDA-approved for food contact, and the liners can withstand temperatures from -40°F to 175°F. Each pack contains 5 liners.
  • Disposable, liquid-tight plastic drum liners
  • Allows complete recovery of wet or dry product from vacuum drums
  • 8 mil low-density polyethylene material
  • Heat-sealed seams for strong, contoured fit
  • Rounded bottom minimizes wrinkles and folds, reducing stress
  • FDA-approved material for contact with food
  • Easy to install; simply twist and tie-off to protect contents
  • Temperature rating from -40°F to 175°F
  • Clear color
Catalog No. 50-283-6338 Supplier MSC Supplier No. 39469721
These disposable drum liners are designed for efficient recovery of wet or dry product from vacuum drums. Made from 8 mil low-density polyethylene, they feature heat-sealed seams and a rounded bottom for a strong, contoured, and wrinkle-free fit. The material is FDA-approved for food contact, and the liners can withstand temperatures from -40°F to 175°F. Each pack contains 5 liners.
  • Disposable, liquid-tight plastic drum liners
  • Allows complete recovery of wet or dry product from vacuum drums
  • 8 mil low-density polyethylene material
  • Heat-sealed seams for strong, contoured fit
  • Rounded bottom minimizes wrinkles and folds, reducing stress
  • FDA-approved material for contact with food
  • Easy to install; simply twist and tie-off to protect contents
  • Temperature rating from -40°F to 175°F
  • Clear color