Learn More
Yamazen Science Inc High Performance Flash Column, Polypropylene, Disposable
Catalog No. NC1465379
Encompass
Universal Columns: Disposable Polypropylene Column with High Throughput Purification Performance, Packed with High Performance Silica GelPore Size 60 angstroms, Particle Size 40-63 microns, Luer Lock TypeSize: 3L
Catalog No. NC1465379 Supplier Yamazen Science Inc Supplier No. UW00806
Universal Columns: Disposable Polypropylene Column with High Throughput Purification Performance, Packed with High Performance Silica GelPore Size 60 angstroms, Particle Size 40-63 microns, Luer Lock TypeSize: 3L