Learn More
Yamazen Science Inc Disposable Column, Packed with High Performance Silica Gel, Luer Lock Type
Catalog No. NC1465374
Encompass
This item has a minimum qty of 2 per supplier requirements.
Universal Columns: Disposable Polypropylene Column with High Throughput Purification Performance, Packed with High Performance Silica GelPore Size 60 angstroms, Particle Size 40-63 microns, Luer Lock TypeSize: L10/pk
Catalog No. NC1465374 Supplier Yamazen Science Inc Supplier No. UW00301
This item has a minimum qty of 2 per supplier requirements.
Universal Columns: Disposable Polypropylene Column with High Throughput Purification Performance, Packed with High Performance Silica GelPore Size 60 angstroms, Particle Size 40-63 microns, Luer Lock TypeSize: L10/pk