Learn More
Micromanipulator DISP PRB NI .35UM TIP (7A)10PK
SDP
Catalog No. NC9417079
Encompass
Probe Tip. Style: Fine Tip, Tip Diameter: 0.005", Point Radius In Microns: 0.350, Material: Tungsten Tip - Nickel Shank, Attributes: Nickel Shank, Bendable Shank Cat-Whisker.
Catalog No. NC9417079 Supplier Micromanipulator Supplier No. 7A
Probe Tip. Style: Fine Tip, Tip Diameter: 0.005", Point Radius In Microns: 0.350, Material: Tungsten Tip - Nickel Shank, Attributes: Nickel Shank, Bendable Shank Cat-Whisker.