Learn More
Micromanipulator DISP PRB TNGSTN .5UM TIP-7B-10
SDP
Catalog No. NC9417084
Encompass
General Purpose Probe Tip 7B 0.020" 0.500 Std Available in custom sizes from 1 to 200 Tungsten General purpose, large target probe tip
Catalog No. NC9417084 Supplier Micromanipulator Supplier No. 7B
General Purpose Probe Tip 7B 0.020" 0.500 Std Available in custom sizes from 1 to 200 Tungsten General purpose, large target probe tip