Learn More
Biopac Systems Inc Disposable RT Dry Electrode, 100/pk
SDP
Catalog No. NC1690154
Encompass
These disposable dry electrodes have a very long shelf-life and are ideal for EDA/GSR applications. EL509 electrodes are radiotranslucent and have circular contact area on an oblong backing. Requires LEAD108B or LEAD108C and electrode gel—GEL101 recommended for EDA/GSR. Standard snap connection. Ag/AgCl laminated, carbon composition contact (11 mm dia). Incorporates gel cavity (16 mm dia 1.5 mm deep). Foam tape backing: 1.5 mm thick, latex free. Strong adhesion. Size: 27 mm x 36 mm
Catalog No. NC1690154 Supplier Biopac Systems Inc Supplier No. EL509
These disposable dry electrodes have a very long shelf-life and are ideal for EDA/GSR applications. EL509 electrodes are radiotranslucent and have circular contact area on an oblong backing. Requires LEAD108B or LEAD108C and electrode gel—GEL101 recommended for EDA/GSR. Standard snap connection. Ag/AgCl laminated, carbon composition contact (11 mm dia). Incorporates gel cavity (16 mm dia 1.5 mm deep). Foam tape backing: 1.5 mm thick, latex free. Strong adhesion. Size: 27 mm x 36 mm