Learn More
DYNAMIC DEVICES DISP TIPS 30UL FOR 384 TPH
Catalog No. NC1699684
Encompass
LXB-384-30NF-1: LM Series, 30μl Disp. Tips, Sterile & Unfiltered Case of LXB Series 30 μl disposable tips, Sterile & Unfiltered For the Lynx Series 384 Tip Pipette Head. Each Case includes 19,200 total tips packaged in 50 racks of 384 tips in a robotic box.
Catalog No. NC1699684 Supplier DYNAMIC DEVICES Supplier No. LXB38430NF1
LXB-384-30NF-1: LM Series, 30μl Disp. Tips, Sterile & Unfiltered Case of LXB Series 30 μl disposable tips, Sterile & Unfiltered For the Lynx Series 384 Tip Pipette Head. Each Case includes 19,200 total tips packaged in 50 racks of 384 tips in a robotic box.