Learn More
Thomas Scientific Tissue Grinder, Large, w / Tube
SDP
Catalog No. NC9617591
Encompass_Preferred
Precision large and small tissue grinder systems Ready-to-use Packaged in a sterile peel pouch Completely disposable Grinder tube doubles as a specimen container for transport and storage. Protective sleeve helps to contain aerosolization created during grinding. Tissue is ground quickly due to fresh grinding surfaces. Specimen can be cooled during grinding.
Catalog No. NC9617591 Supplier Thomas Scientific Supplier No. 3432N95
Precision large and small tissue grinder systems Ready-to-use Packaged in a sterile peel pouch Completely disposable Grinder tube doubles as a specimen container for transport and storage. Protective sleeve helps to contain aerosolization created during grinding. Tissue is ground quickly due to fresh grinding surfaces. Specimen can be cooled during grinding.