Learn More
Andwin Scientific DISP.TRANSFER PIPET,STL,500/PK
SDP
Catalog No. NC0362007
Encompass_Preferred
The disposable 7.5 mL transfer pipet is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding. This product is sold in a pack of 500.
Catalog No. NC0362007 Supplier Andwin Scientific Supplier No. 414004005
The disposable 7.5 mL transfer pipet is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding. This product is sold in a pack of 500.