Learn More
Chemglass Life Sciences Tubing Set with Pump Head
SDP
Catalog No. 501532101
Encompass
Tubing Set with Pump Head. Tubing Complete Set includes pre-assembled tubes, reusable pump head and head plate with a sampling port to support the continuous culture in BelloCell-500P system. It can be autoclaved at 121 C for 30 minutes. Tubing Set (CLS-1328-01) Includes (1) Each: Disposable Tubing Kit Pump Head
Catalog No. 50-153-2101 Supplier Chemglass Life Sciences Supplier No. CLS132801
Tubing Set with Pump Head. Tubing Complete Set includes pre-assembled tubes, reusable pump head and head plate with a sampling port to support the continuous culture in BelloCell-500P system. It can be autoclaved at 121 C for 30 minutes. Tubing Set (CLS-1328-01) Includes (1) Each: Disposable Tubing Kit Pump Head