Learn More
Med Vet International Disp Vet Warming Blanket 67" X 39" Med Over Body, J1534, Each
Catalog No. 5000583149
Encompass_Preferred
Disposable Veterinary Warming Blanket. The blankets are designed for single use, but with care they can be re-used to some extent. Clean with a damp towel or reuse them by drying the blanket by connecting to warm air unit. • Use tie to attach to the hose. J1534 Blanket Type Medium Over. Dimensions 67" x 39" (170cm x 100cm)
Catalog No. 50-005-83149 Supplier Med Vet International Supplier No. JOR1534
Disposable Veterinary Warming Blanket. The blankets are designed for single use, but with care they can be re-used to some extent. Clean with a damp towel or reuse them by drying the blanket by connecting to warm air unit. • Use tie to attach to the hose. J1534 Blanket Type Medium Over. Dimensions 67" x 39" (170cm x 100cm)