Learn More
Belair Instrument Company Inc Avantik Square Edge Trim Blade Dispenser, Touch-Free Loading, 130 mm Blade Length, 25 Blades Per Pack, Unit
SDP
Supplier:  Belair Instrument Company Inc GR2143A
Encompass_Preferred
The Avantik Square Edge Trim Blade Dispenser is designed for precise tissue sectioning in labs, offering a safe, touch-free blade loading system. It dispenses a pack of 25 square edge blades, enhancing workflow efficiency with quick loading while minimizing direct contact with blades. Ideal for histology and pathology, this dispenser ensures user safety and convenience, setting it apart from typical blade dispensers.
Catalog No. NC1282891