Learn More
STEMCELL Technologies Dispase (1 U/mL), Size: 100 mL
SDP
Catalog No. NC9995391
Encompass
Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues. Incubation of minced tissue with pre-warmed dispase and gentle agitation will liberate cells with minimal cell damage. Pre-warmed dispase can also be used to harvest cells from tissue culture plastic. Unlike trypsin, dispase is not inhibited by serum. Dispase activity is inhibited by EDTA and EGTA. Dispase should be removed from cell suspensions by centrifuging the cells followed by washing the cells with buffer or culture medium.
Catalog No. NC9995391 Supplier STEMCELL Technologies Supplier No. 07923
Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues. Incubation of minced tissue with pre-warmed dispase and gentle agitation will liberate cells with minimal cell damage. Pre-warmed dispase can also be used to harvest cells from tissue culture plastic. Unlike trypsin, dispase is not inhibited by serum. Dispase activity is inhibited by EDTA and EGTA. Dispase should be removed from cell suspensions by centrifuging the cells followed by washing the cells with buffer or culture medium.