Learn More
STEMCELL Technologies Dispase (5 U/mL), Size: 100 mL
SDP
Catalog No. NC9886504
Encompass
Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues. This product contains 5 U/mL Dispase II (neutral protease from Bacillus polymyxa) dissolved in Hanks’ Balanced Salt Solution.
Catalog No. NC9886504 Supplier STEMCELL Technologies Supplier No. 07913
Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues. This product contains 5 U/mL Dispase II (neutral protease from Bacillus polymyxa) dissolved in Hanks’ Balanced Salt Solution.