Learn More
Sigma Aldrich Fine Chemicals Biosciences Dispase II (neutral protease, grade II), 5 x 1 g
Catalog No. NC1137081
Encompass
Dispase is used for the preparation of cells from a wide variety of different tissues and organs. The enzyme has proven to be a rapid, effective, yet gentle agent for the disruption of extracellular matrix of tissues for releasing single cells for cell culture (primary cell culture) or harvesting cells already in culture to transfer to new substrate (secondary culture). Furthermore, it is used to prevent unwanted clumping of cells cultured in suspension.
Catalog No. NC1137081 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4942078001
Dispase is used for the preparation of cells from a wide variety of different tissues and organs. The enzyme has proven to be a rapid, effective, yet gentle agent for the disruption of extracellular matrix of tissues for releasing single cells for cell culture (primary cell culture) or harvesting cells already in culture to transfer to new substrate (secondary culture). Furthermore, it is used to prevent unwanted clumping of cells cultured in suspension.