Learn More
Electron Microscopy Sciences Adjustable Volume LDPE Dispenser Bottle 1000 ml, 5 to 25 ml
SDP
Catalog No. 501928296
Encompass_Preferred
These adjustable volume bottles have a graduated clear PMP measuring cup that slides up and down on the vertical polyethylene tube for easy adjustment. The variation in height determines the volume to be dispensed. The cup is filled by squeezing the bottle and forcing the liquid up the tube. When the bottle pressure is released, the excess will be drawn back into the bottle and only the desired volume will remain.
Gradations: 5 to 25 ml
Catalog No. 50-192-8296 Supplier Electron Microscopy Sciences Supplier No. 6429703
These adjustable volume bottles have a graduated clear PMP measuring cup that slides up and down on the vertical polyethylene tube for easy adjustment. The variation in height determines the volume to be dispensed. The cup is filled by squeezing the bottle and forcing the liquid up the tube. When the bottle pressure is released, the excess will be drawn back into the bottle and only the desired volume will remain.
Gradations: 5 to 25 ml