Learn More
Thomas Scientific Metal Dispenser for 10 x 75mm and 12 x 75mm glass culture tubes
SDP
Catalog No. 501461435
Encompass
Globe Scientific is pleased to offer these premium quality borosilicate glass tubes. The tubes provide excellent chemical resistance and durability. They feature well-rounded bottoms and smooth, fire-polished rims. Dimensional specifications are rigidly controlled for reliable, uniform results. The tubes are produced in an ISO 9001:2008 certified factory. They are available in six sizes. Tubes can be used for tissue culture and blood banks Made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from tubes made from soda lime glass Tubes feature well-rounded bottoms and smooth fire-polished rims Our exclusive TubeSaver™ Packaging System helps prevent breakage and contamination during shipping and storage. The tubes are neatly stacked in heavy-duty, shrink wrapped boxes of 250 tubes, 4 boxes per case. For additional convenience, the 10 x 75mm, 12 x 75mm and 13 x 100mm boxes fit into Globe Scientific’s metal/plexiglass t
Catalog No. 50-146-1435 Supplier Thomas Scientific Supplier No. 1218M75
Globe Scientific is pleased to offer these premium quality borosilicate glass tubes. The tubes provide excellent chemical resistance and durability. They feature well-rounded bottoms and smooth, fire-polished rims. Dimensional specifications are rigidly controlled for reliable, uniform results. The tubes are produced in an ISO 9001:2008 certified factory. They are available in six sizes. Tubes can be used for tissue culture and blood banks Made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from tubes made from soda lime glass Tubes feature well-rounded bottoms and smooth fire-polished rims Our exclusive TubeSaver™ Packaging System helps prevent breakage and contamination during shipping and storage. The tubes are neatly stacked in heavy-duty, shrink wrapped boxes of 250 tubes, 4 boxes per case. For additional convenience, the 10 x 75mm, 12 x 75mm and 13 x 100mm boxes fit into Globe Scientific’s metal/plexiglass t