Learn More
Cole-Parmer Thermo Scientific 4421150 Finnpipette(R) Dispenser, 2.0 to 10.0 mL
Catalog No. 50200187 Shop All Cole-Parmer Products
Encompass
These dispensers offer simple, rapid dispensing with reliable results through the unique combination of ceramic piston, graduated gauge, and digital volumetric adjustment device. This design provides absolute repoducibility and is calibrated for life. Dispensers are suitable for A32 thread.
Catalog No. 50-200-187 Supplier Cole-Parmer Supplier No. 0790345
These dispensers offer simple, rapid dispensing with reliable results through the unique combination of ceramic piston, graduated gauge, and digital volumetric adjustment device. This design provides absolute repoducibility and is calibrated for life. Dispensers are suitable for A32 thread.