Learn More
HF Scientific Inc Total Chlorine DPD Reagent Dispenser, 100 Tests, 5ml
Catalog No. NC1748988
Encompass_Preferred
This DPD reagent dispenser provides for 100 tests for total chlorine. It has a 5ml sample size, for use with HF scientific and Hach photometers.
Catalog No. NC1748988 Supplier HF Scientific Inc Supplier No. 10502
This DPD reagent dispenser provides for 100 tests for total chlorine. It has a 5ml sample size, for use with HF scientific and Hach photometers.